N-Boc-D-serine methyl ester 98%

Supplier: Ambeed
Warning

Synonyms: N-(tert-Butoxycarbonyl)-D-serine methyll ester, Boc-D-Ser-OMe, N-Boc-D-serine methyll ester

A452649-100G A452649-10G A452649-500G A452649-1000G A452649-1G A452649-5G
103810-358EA 297 USD
103810-358 103827-806 103838-876 76877-524 76877-526 76877-528 77307-370 77307-372 77307-374
N-Boc-D-serine methyl ester 98%
N-Boc-D-serine methyl ester

Precaución: for Research Use Only

Formula: C₉H₁₇NO₅
MW: 219.237 g/mol
Boiling Pt: 354.3 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00270516
CAS Number: 95715-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR