N-(tert-Butoxycarbonyl)-D-serine 98%

Supplier: Ambeed
Warning

Synonyms: N-Boc-D-serine, Boc-D-Ser-OH

A135826-100G A135826-10G A135826-500G A135826-25G A135826-5G
76811-498EA 47.75 USD
76811-498 76811-500 76811-502 77315-088 77315-090
N-(tert-Butoxycarbonyl)-D-serine 98%
N-(tert-Butoxycarbonyl)-D-serine

Precaución: for Research Use Only

Formula: C₈H₁₅NO₅
MW: 205.21 g/mol
Boiling Pt: 385.1 °C (760 mm Hg)
Melting Pt: 91…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00063142
CAS Number: 6368-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR