Boc-D-Serinol(Bzl) 97%

Supplier: Ambeed
Warning

A325274-100MG A325274-250MG A325274-1G
103825-396EA 106 USD
103825-396 103836-338 76693-830 76693-832 77306-326
Boc-D-Serinol(Bzl) 97%
Boc-D-Serinol(Bzl)

Precaución: for Research Use Only

Formula: C₁₅H₂₃NO₄
MW: 281.3 g/mol
Boiling Pt: 443.099 °C (760 mm Hg)
Melting Pt: 66…69 °C (760 mm Hg)
Flash Pt: 221.779 °C
Storage Temperature: Ambient
MDL Number: MFCD00235944
CAS Number: 127559-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR