O-Benzyl-N-(tert-butoxycarbonyl)-D-serine 97%

Supplier: Ambeed
Warning

A326243-1G A326243-25G A326243-5G A326243-100G A326243-10G A326243-500G
103825-422EA 5 USD
103825-422 103836-370 76839-318 76839-320 76839-322 76978-164 76978-166 77881-912
O-Benzyl-N-(tert-butoxycarbonyl)-D-serine 97%
O-Benzyl-N-(tert-butoxycarbonyl)-D-serine

Precaución: For research use only

Formula: C₁₅H₂₁NO₅
Boiling Pt: 456.2 °C (760 mm Hg)
Melting Pt: 58…60 °C
Flash Pt: 229.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00038248
CAS Number: 47173-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR