Boc-D-Pro-OH 97%

Supplier: Ambeed
Warning

Synonyms: (2R)-1-[(tert-butoxy)carbonyl]pyrrolidine-2-carboxylic acid, N-Boc-D-proline

A453679-100G A453679-10G A453679-500G A453679-25G A453679-5G A453679-1000G
103838-886EA 5 USD
103838-886 76695-258 76695-260 76695-262 76842-720 76842-722 77626-898
Boc-D-Pro-OH 97%
Boc-D-Pro-OH

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₄
MW: 215.25 g/mol
Boiling Pt: 337.2 °C (760 mm Hg)
Melting Pt: 134…137 °C
Flash Pt: 157.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00063226
CAS Number: 37784-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR