N-(tert-Butoxycarbonyl)-D-phenylalanine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Boc-D-phenylalanine, BOC-D-Phenylalanine, Boc-D-Phe-OH
A555031-25G
A555031-5G
A555031-100G
A555031-10G
A555031-1G
A555031-500G
103839-840EA
6
USD
103839-840
76843-928
76843-930
77197-538
77197-540
77197-542
77197-544
N-(tert-Butoxycarbonyl)-D-phenylalanine 98%
N-(tert-Butoxycarbonyl)-D-phenylalanine
Precaución: for Research Use Only
Formula:
C₁₄H₁₉NO₄ MW: 265.31 g/mol Boiling Pt: 426.6 °C (760 mm Hg) Melting Pt: 78…87 °C Flash Pt: 211.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063149 CAS Number: 18942-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...