(R)-N-(tert-Butoxycarbonyl)-2-phenylglycinol 97%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Phenylglycinol, (-)-N-Boc-D-α-phenylglycinol

A227101-100G A227101-10G A227101-500G A227101-1G A227101-25G A227101-5G
103819-296EA 5 USD
103819-296 103850-202 77013-412 77013-414 77013-416 77088-956 77088-958 77088-960
(R)-N-(tert-Butoxycarbonyl)-2-phenylglycinol 97%
(R)-N-(tert-Butoxycarbonyl)-2-phenylglycinol

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO₃
Melting Pt: 137…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00274205
CAS Number: 102089-74-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR