Boc-4-methyl-D-phenylalanine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-D-Phe(4-Me)-OH, Boc-p-Me-D-Phe-OH
A591842-1G
A591842-5G
A591842-10G
103828-930EA
48
USD
103828-930
103840-076
76844-222
76844-224
76878-896
Boc-4-methyl-D-phenylalanine 98%
Boc-4-methyl-D-phenylalanine
Precaución: for Research Use Only
Formula:
C₁₅H₂₁NO₄ MW: 279.336 g/mol Boiling Pt: 440.4 °C (760 mm Hg) Melting Pt: 85 °C (760 mm Hg) Flash Pt: 220.2 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01317726 CAS Number: 80102-27-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...