Boc-D-4-iodophenylalanine 95%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Phe(4-I)-OH, N-Boc-4-iodo-D-phenylalanine, Boc-p-iodo-D-Phe-OH

A130936-10G A130936-250MG A130936-25G A130936-5G
103801-496EA 7 USD
103801-496 76782-332 76782-334 77313-976 77313-978
Boc-D-4-iodophenylalanine 95%
Boc-D-4-iodophenylalanine

Precaución: for Research Use Only

Formula: C₁₄H₁₈INO₄
MW: 391.205 g/mol
Boiling Pt: 475.3 °C (760 mm Hg)
Flash Pt: 241.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00079670
CAS Number: 176199-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR