N-(tert-Butoxycarbonyl)-4-fluoro-D-phenylalanine 95%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Phe(4-F)-OH, N-Boc-4-fluoro-D-phenylalanine

A105485-250MG A105485-1G
103796-890EA 22 USD
103796-890 103822-622 77188-762 77309-490
N-(tert-Butoxycarbonyl)-4-fluoro-D-phenylalanine 95%
N-(tert-Butoxycarbonyl)-4-fluoro-D-phenylalanine

Precaución: for Research Use Only

Formula: C₁₄H₁₈FNO₄
MW: 283.30 g/mol
Boiling Pt: 431.2 °C (760 mm Hg)
Melting Pt: 83…86 °C
Flash Pt: 214.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00079669
CAS Number: 57292-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR