(R)-N-Boc-3-fluorophenylalanine 95%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Phe(3-F)-OH

A130013-100G A130013-100MG A130013-10G A130013-250MG A130013-1G A130013-25G
103801-366EA 28 USD
103801-366 76996-332 76996-334 76996-336 76996-338 77313-792 77313-794
(R)-N-Boc-3-fluorophenylalanine 95%
(R)-N-Boc-3-fluorophenylalanine

Precaución: for Research Use Only

Formula: C₁₄H₁₈FNO₄
MW: 283.3 g/mol
Boiling Pt: 425 °C (760 mm Hg)
Melting Pt: 75…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00672523
CAS Number: 114873-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR