Boc-D-Lys(fmoc)-OH 95%

Supplier: Ambeed
Warning

Synonyms: Nα-Boc-Nepsilon-Fmoc-D-lysine, Boc-D-Lys(Fmoc)-OH

A214392-10G A214392-250MG A214392-1G A214392-5G
103818-394EA 16 USD
103818-394 103849-100 76999-714 76999-716 77087-618 77303-342
Boc-D-Lys(fmoc)-OH 95%
Boc-D-Lys(fmoc)-OH

Precaución: for Research Use Only

Formula: C₂₆H₃₂N₂O₆
MW: 468.55 g/mol
Boiling Pt: 685.7 °C (760 mm Hg)
Melting Pt: 117…123 °C
Flash Pt: 368.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00076966
CAS Number: 115186-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR