Boc-DL-Pro-OH 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(tert-Butoxycarbonyl)pyrrolidine-2-carboxylic acid, N-Boc-DL-proline
A219810-100G
A219810-10G
A219810-1G
A219810-500G
A219810-25G
A219810-5G
103849-698EA
6
USD
103849-698
76691-768
76691-770
76691-772
76691-774
77088-264
77088-266
Boc-DL-Pro-OH 97%
Boc-DL-Pro-OH
Precaución: for Research Use Only
Formula:
C₁₀H₁₇NO₄ Boiling Pt: 337.2 °C (760 mm Hg) Melting Pt: 133…135 °C Flash Pt: 157.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00520915 CAS Number: 59433-50-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...