N-(tert-Butoxycarbonyl)iminodiacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: Boc-Ida-OH, Boc-DL-Ida-OH, N-Boc-iminodiacetic Acid

A217835-100G A217835-500G A217835-1G A217835-25G
103818-662EA 6 USD
103818-662 103849-390 76691-680 76691-682 77088-032 77303-466
N-(tert-Butoxycarbonyl)iminodiacetic acid 97%
N-(tert-Butoxycarbonyl)iminodiacetic acid

Precaución: for Research Use Only

Formula: C₉H₁₅NO₆
MW: 233.22 g/mol
Boiling Pt: 412 °C (760 mm Hg)
Flash Pt: 203 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00800351
CAS Number: 56074-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR