N-(tert-Butoxycarbonyl)-D-leucine hydrate 95%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Boc-D-leucine, Boc-D-Leu-OH
A323553-1G
A323553-500G
A323553-100G
A323553-10G
103825-344EA
6
USD
103825-344
103836-284
77306-304
77683-962
77892-666
77892-667
N-(tert-Butoxycarbonyl)-D-leucine hydrate 95%, Ambeed.Inc
N-(tert-Butoxycarbonyl)-D-leucine hydrate
Precaución: For research use only.
Formula:
C₁₁H₂₁NO₄ MW: 231.29 g/mol Boiling Pt: 356 °C (760 mm Hg) Melting Pt: 85…87 °C Flash Pt: 169.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00038294 CAS Number: 16937-99-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...