2-((tert-Butoxycarbonyl)amino)propanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: N-Boc-DL-alanine, N-(tert-Butoxycarbonyl)-DL-alanine, Boc-DL-Ala-OH

A160646-10G A160646-5G A160646-25G A160646-100G
103808-888EA 320 USD
103808-888 103843-634 77191-070 77300-478 77319-584 77746-097
2-((tert-Butoxycarbonyl)amino)propanoic acid 95%
2-((tert-Butoxycarbonyl)amino)propanoic acid

Precaución: Keep in a dark place, sealed in dry, room temperature

Formula: C₈H₁₅NO₄
MW: 189.2 g/mol
Boiling Pt: 320.9 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00063124
CAS Number: 3744-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR