2-[(tert-Butoxycarbonyl)amino]butanoic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-DL-Abu-OH, 2-Aminobutanoic acid, N-BOC protected
A145429-250MG
A145429-1G
A145429-25G
A145429-5G
A145429-10G
A145429-100G
103803-532EA
9
USD
103803-532
103841-618
76689-146
77316-906
77316-908
77316-910
77624-204
77876-989
2-[(tert-Butoxycarbonyl)amino]butanoic acid 95%
2-[(tert-Butoxycarbonyl)amino]butanoic acid
Precaución: for Research Use Only
Formula:
C₉H₁₇NO₄ MW: 203.24 g/mol Boiling Pt: 334.5±25.0 °C (760 mm Hg) Flash Pt: 156.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00270358 CAS Number: 77284-64-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...