(R)-1-Boc-piperidine-2-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-(+)-N-BOC-2-Piperidinecarboxylic acid, Boc-D-Homopro-OH, Boc-D-HoPro-OH, (R)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid
A163702-100G
A163702-10G
A163702-25G
A163702-1G
A163702-5G
A163702-500G
103805-748EA
9
USD
103805-748
103844-010
76799-104
76799-106
77300-668
77320-118
77320-120
77667-290
(R)-1-Boc-piperidine-2-carboxylic acid 95%
(R)-1-Boc-piperidine-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₁H₁₉NO₄ Melting Pt: 116…119 °C Storage Temperature: Ambient |
MDL Number:
MFCD00237380 CAS Number: 28697-17-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...