Boc-D-homophenylalanine 98%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Homophe-OH, (R)-2-(Boc-amino)-4-phenylbutyric acid, Boc-D-HoPhe-OH

A404968-100G A404968-10G A404968-250MG A404968-1G A404968-5G A404968-25G
103827-112EA 17 USD
103827-112 103838-154 76788-978 76788-980 76788-982 76841-640 76841-642 77307-094
Boc-D-homophenylalanine 98%
Boc-D-homophenylalanine

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₄
MW: 279.336 g/mol
Boiling Pt: 439.6±38.0 °C (760 mm Hg)
Melting Pt: 71…78 °C
Flash Pt: 219.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00076905
CAS Number: 82732-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR