Boc-D-glutamine 98%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Gln-OH

A101040-250MG A101040-100G A101040-25G A101040-10G
103795-774EA 8 USD
103795-774 103821-464 77592-064 77599-474 77599-476 77623-818
Boc-D-glutamine 98%
Boc-D-glutamine

Precaución: Keep in a dark place, sealed in dry, room temperature

Formula: C₁₀H₁₈N₂O₅
MW: 246.263 g/mol
Boiling Pt: 509.1 °C (760 mm Hg)
Melting Pt: 117…119 °C
Flash Pt: 261.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00038158
CAS Number: 61348-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR