Boc-D-Chg-OH 97%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Chg-OH, N-(tert-Butoxycarbonyl)-D-2-cyclohexylglycine, N-Boc-2-cyclohexyl-D-glycine, Boc-cyclohexyl-D-Gly-OH, (R)-2-((tert-Butoxycarbonyl)amino)-2-cyclohexylacetic acid, BOC-D-α-cyclohexylglycine, N-Boc-D-2-cyclohexylglycine

A126563-10G A126563-250MG A126563-1G A126563-25G A126563-5G A126563-100G A126563-500G
103800-834EA 6 USD
103800-834 76718-752 76718-754 77313-056 77313-058 77313-060 77666-804 77666-806
Boc-D-Chg-OH 97%
Boc-D-Chg-OH

Precaución: for Research Use Only

Formula: C₁₃H₂₃NO₄
MW: 257.33 g/mol
Flash Pt: 200.5 °C
Storage Temperature: Freezer
MDL Number: MFCD00133629
CAS Number: 70491-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR