N-Boc-D-aspartic acid-1-methyl ester 97%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Asp-OMe, (R)-3-(Boc-amino)-4-methoxy-4-oxobutyric acid

A193320-1G A193320-5G A193320-100G A193320-10G A193320-25G
103816-522EA 5 USD
103816-522 103847-136 77085-034 77085-036 77151-944 77151-946 77302-336
N-Boc-D-aspartic acid-1-methyl ester 97%
N-Boc-D-aspartic acid-1-methyl ester

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₆
Boiling Pt: 407.1±40.0 °C (760 mm Hg)
Flash Pt: 200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094776
CAS Number: 137130-65-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR