Boc-D-Asp(OMe)-OH 95%

Supplier: Ambeed
Warning

A124190-10G A124190-250MG A124190-25G A124190-1G A124190-5G
103800-496EA 6 USD
103800-496 77026-676 77026-678 77026-680 77312-626 77312-628
Boc-D-Asp(OMe)-OH 95%
Boc-D-Asp(OMe)-OH

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₆
MW: 247.25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11617949
CAS Number: 124184-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR