Nɑ-(tert-Butoxycarbonyl)-D-asparagine 97%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Asn-OH, Nɑ-Boc-D-asparagine

A354482-25G A354482-100G A354482-10G A354482-500G A354482-1G A354482-5G
103826-124EA 5 USD
103826-124 103837-128 76840-250 77263-902 77263-904 77263-906 77306-664 77306-666
Nɑ-(tert-Butoxycarbonyl)-D-asparagine 97%
Nalpha-(tert-Butoxycarbonyl)-D-asparagine

Precaución: for Research Use Only

Formula: C₉H₁₆N₂O₅
MW: 232.24 g/mol
Boiling Pt: 481.7±40.0 °C (760 mm Hg)
Melting Pt: 165…169 °C
Flash Pt: 245.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00065558
CAS Number: 75647-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR