(S)-3-Amino-2-(tert-butoxycarbonylamino)propionic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-N-Boc-L-alanine, Nalpha-Boc-2,3-diaminopropionic Acid, Boc-Dpr-OH, Boc-Dap-OH, 3-Amino-N-(tert-butoxycarbonyl)-L-alanine

A100715-100G A100715-10G A100715-25G A100715-1G A100715-5G
103795-590EA 5 USD
103795-590 103821-288 77107-160 77107-162 77107-164 77282-548 77296-828
(S)-3-Amino-2-(tert-butoxycarbonylamino)propionic acid 97%
(S)-3-Amino-2-(tert-butoxycarbonylamino)propionic acid

Precaución: for Research Use Only

Formula: C₈H₁₆N₂O₄
MW: 204.23 g/mol
Boiling Pt: 364.4 °C (760 mm Hg)
Melting Pt: 210 °C
Flash Pt: 174.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00236843
CAS Number: 73259-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR