Boc-Dap(Cbz)-OH 97%

Supplier: Ambeed
Warning

Synonyms: Boc-Dap(Z)-OH, (S)-2-[(tert-Butoxycarbonyl)amino]-3-(benzyloxycarbonylamino)propionic acid

A253941-10G A253941-250MG A253941-25G A253941-1G A253941-5G A253941-100G
103812-116EA 6 USD
103812-116 103851-968 76692-574 76692-576 76692-578 77091-406 77304-622 77626-332
Boc-Dap(Cbz)-OH 97%
Boc-Dap(Cbz)-OH

Precaución: for Research Use Only

Formula: C₁₆H₂₂N₂O₆
MW: 338.36 g/mol
Melting Pt: 97…100 °C
Flash Pt: 285.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08275856
CAS Number: 65710-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR