Boc-D-Ala-NH2 97%

Supplier: Ambeed
Warning

A101029-100G A101029-10G A101029-250MG A101029-25G A101029-1G
103795-764EA 6 USD
103795-764 103821-456 76687-014 76687-016 76687-018 76687-020 77296-958
Boc-D-Ala-NH2 97%
Boc-D-Ala-NH2

Precaución: for Research Use Only

Formula: C₈H₁₆N₂O₃
MW: 188.2 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03701448
CAS Number: 78981-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR