(S)-4-Amino-2-(tert-butoxycarbonylamino)butyric acid 95%

Supplier: Ambeed
Danger

Synonyms: Boc-Dab-OH, (S)-4-Amino-2-(Boc-amino)butyric Acid, (S)-4-Amino-2-(tert-butoxycarbonylamino)butyric Acid, (S)-Nalpha-Boc-2,4-diaminobutyric Acid, (S)-4-Amino-2-(Boc-amino)butyric acid (S)-Nα-Boc-2,4-diaminobutyric acid Boc-Dab-OH

A112589-10G A112589-250MG A112589-1G A112589-5G A112589-25G A112589-100G
103798-048EA 16 USD
103798-048 103823-886 77094-070 77094-072 77310-696 77310-698 77570-694 77684-999
(S)-4-Amino-2-(tert-butoxycarbonylamino)butyric acid 95%
(S)-4-Amino-2-(tert-butoxycarbonylamino)butyric acid

Precaución: for Research Use Only

Formula: C₉H₁₈N₂O₄
Boiling Pt: 385.5 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00236841
CAS Number: 25691-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR