4-Bromo-N-(3-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide 95%

Supplier: Ambeed
Warning

Synonyms: Boc-D-3-Abu-OH

A136124-100G A136124-25G A136124-5G A136124-10G A136124-500G
103802-276EA 5 USD
103802-276 77137-090 77137-092 77315-172 77745-966 77745-967
4-Bromo-N-(3-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide 95%
4-Bromo-N-(3-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide

Precaución: for Research Use Only

Formula: C₉H₁₇NO₄
MW: 203.238 g/mol
Boiling Pt: 339.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00270343
CAS Number: 159991-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR