N-(tert-Butoxycarbonyl)-3-(1-naphthyl)-D-alanine 97%

Supplier: Ambeed
Warning

Synonyms: (R)-alpha-tert-Butoxycarbonylamino-1-naphthalenepropionic Acid, Boc-D-1-Nal-OH, (R)-alpha-(Boc-amino)-1-naphthalenepropionic Acid, N-Boc-3-(1-naphthyl)-D-alanine, (R)-2-((tert-Butoxycarbonyl)amino)-3-(naphthalen-1-yl)propanoic acid

A123923-250MG A123923-25G A123923-1G A123923-5G
103800-466EA 39 USD
103800-466 77026-664 77026-666 77312-594 77312-596
N-(tert-Butoxycarbonyl)-3-(1-naphthyl)-D-alanine 97%
N-(tert-Butoxycarbonyl)-3-(1-naphthyl)-D-alanine

Precaución: for Research Use Only

Formula: C₁₈H₂₁NO₄
MW: 315.37 g/mol
Boiling Pt: 512.2 °C (760 mm Hg)
Melting Pt: 145…148 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00079668
CAS Number: 76932-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR