N-Boc-S-trityl-L-cysteine 98%

Supplier: Ambeed
Warning

Synonyms: N-Boc-S-trityl-L-cysteine, N-(tert-Butoxycarbonyl)-S-trityl-L-cysteine, BOC-S-trityl-L-cysteine, (R)-2-((tert-Butoxycarbonyl)amino)-3-(tritylthio)propanoic acid, Boc-Cys(Trt)-OH

A147187-100G A147187-10G A147187-500G A147187-25G A147187-5G
103841-844EA 9 USD
103841-844 76997-154 76997-156 76997-158 77317-210 77317-212
N-Boc-S-trityl-L-cysteine 98%
N-Boc-S-trityl-L-cysteine

Precaución: for Research Use Only

Formula: C₂₇H₂₉NO₄S
Boiling Pt: 610.9 °C (760 mm Hg)
Melting Pt: 143…146 °C
Flash Pt: 323.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00038251
CAS Number: 21947-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR