Boc-cycloleucine 95%

Supplier: Ambeed
Warning

Synonyms: 1-(Boc-amino)cyclopentanecarboxylic acid, 1-N-BOC-Aminocyclopentanecarboxylic acid, 1-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid

A274768-10G A274768-1G
103813-218EA 9 USD
103813-218 103853-146 77099-066 77305-172
Boc-cycloleucine 95%
Boc-cycloleucine

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
Boiling Pt: 376.6 °C (760 mm Hg)
Melting Pt: 130…135 °C
Storage Temperature: Ambient
MDL Number: MFCD01076126
CAS Number: 35264-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR