(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclohexylacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: N-Boc-2-cyclohexyl-L-glycine, Boc-cyclohexyl-Gly-OH, BOC-L-α-cyclohexylglycine, N-Boc-L-2-cyclohexylglycine, Boc-Chg-OH

A104896-100G A104896-10G A104896-250MG A104896-25G A104896-500G A104896-1G A104896-5G
103796-794EA 6 USD
103796-794 103822-520 76994-788 76994-790 76994-792 76994-794 76994-796 77309-382 77309-384
(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclohexylacetic acid 97%
(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclohexylacetic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₃NO₄
MW: 257.3 g/mol
Boiling Pt: 407.9 °C (760 mm Hg)
Melting Pt: 72…76 °C
Flash Pt: 200.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00133638
CAS Number: 109183-71-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR