Boc-Asp-OtBu 97%

Supplier: Ambeed
Warning

Synonyms: Boc-Asp-OtBu, N-Boc-L-aspartic acid 1-tert-Butyl ester, N-(tert-Butoxycarbonyl)-L-aspartic acid 1-tert-Butyl ester, 1-tert-Butyl N-Boc-L-aspartate, 1-tert-Butyl N-(tert-Butoxycarbonyl)-L-aspartate, N-Boc-L-aspartic acid 1-tert-butyl ester

A240695-1G A240695-100G A240695-10G A240695-500G A240695-25G A240695-5G
103811-356EA 8 USD
103811-356 103851-160 77090-314 77165-762 77165-764 77165-766 77304-196 77304-198
Boc-Asp-OtBu 97%
Boc-Asp-OtBu

Precaución: for Research Use Only

Formula: C₁₃H₂₃NO₆
MW: 289.328 g/mol
Boiling Pt: 429 °C (760 mm Hg)
Melting Pt: 101…103 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00038272
CAS Number: 34582-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR