1-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate 98%

Supplier: Ambeed
Warning

Synonyms: 1-Benzyl N-Boc-L-aspartate, N-(tert-Butoxycarbonyl)-L-aspartic Acid 1-Benzyl Ester, BOC-L-Aspartic acid 1-benzyl ester, N-Boc-L-aspartic Acid 1-Benzyl Ester, Boc-Asp-OBzl, Boc-Asp-Obzl

A256519-5G A256519-100G A256519-10G A256519-25G A256519-500G
103852-136EA 9 USD
103852-136 77091-632 77276-446 77276-448 77304-680 77748-479
1-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate 98%
1-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate

Precaución: for Research Use Only

Formula: C₁₆H₂₁NO₆
MW: 323.35 g/mol
Boiling Pt: 504.3 °C (760 mm Hg)
Melting Pt: 95…97 °C
Flash Pt: 258.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00065563
CAS Number: 30925-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR