4-tert-Butyl-N-(tert-butoxycarbonyl)-L-aspartate 97%

Supplier: Ambeed
Warning

Synonyms: 4-tert-Butyl N-Boc-L-aspartate, N-(tert-Butoxycarbonyl)-L-aspartic Acid 4-tert-Butyl Ester, N-Boc-L-aspartic Acid 4-tert-Butyl Ester, Boc-Asp(OtBu)-OH

A148662-100G A148662-10G A148662-1G A148662-5G
103803-954EA 5 USD
103803-954 103842-070 76997-252 76997-254 77299-676 77317-532
4-tert-Butyl-N-(tert-butoxycarbonyl)-L-aspartate 97%
4-tert-Butyl-N-(tert-butoxycarbonyl)-L-aspartate

Precaución: for Research Use Only

Formula: C₁₃H₂₃NO₆
MW: 289.33 g/mol
Melting Pt: 64…67 °C
Flash Pt: 215.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00076912
CAS Number: 1676-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR