N-ɑ-Boc-L-asparagine 98%

Supplier: Ambeed
Warning

Synonyms: Boc-L-Asparagine, Boc-Asn-OH

A105411-100G A105411-10G A105411-25G A105411-500G
103807-930EA 263 USD
103807-930 77270-478 77270-480 77270-482 77270-484
N-ɑ-Boc-L-asparagine 98%
N-alpha-Boc-L-asparagine

Precaución: for Research Use Only

Formula: C₉H₁₆N₂O₅
MW: 232.24 g/mol
Boiling Pt: 481.7 °C (760 mm Hg)
Melting Pt: 175 °C
Flash Pt: 245.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00038152
CAS Number: 7536-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR