[(tert-Butoxycarbonyl)aminooxy]acetic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Boc-(carboxymethoxy)amine, Boc-Aoa-OH, (Boc-aminooxy)acetic Acid
A237626-1G
A237626-100G
A237626-10G
A237626-25G
A237626-5G
103811-166EA
6
USD
103811-166
103850-946
77090-040
77140-188
77140-190
77304-086
77304-088
[(tert-Butoxycarbonyl)aminooxy]acetic acid 98%
[(tert-Butoxycarbonyl)aminooxy]acetic acid
Precaución: for Research Use Only
Formula:
C₇H₁₃NO₅ MW: 191.18 g/mol Melting Pt: 115 °C Storage Temperature: Ambient |
MDL Number:
MFCD01632027 CAS Number: 42989-85-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...