(S)-N-tert-Butoxycarbonylalanine methyl ester 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-Ala-OMe, N-Boc-L-alanine methyl ester
A329488-10G
A329488-5G
A329488-100G
103836-482EA
5
USD
103836-482
77003-008
77306-396
77672-694
(S)-N-tert-Butoxycarbonylalanine methyl ester 95%
(S)-N-tert-Butoxycarbonylalanine methyl ester
Precaución: for Research Use Only
Formula:
C₉H₁₇NO₄ Boiling Pt: 277.8 °C (760 mm Hg) Melting Pt: 32…35 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00038513 CAS Number: 28875-17-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...