N-(tert-Butoxycarbonyl)-L-alanine N'-methoxy-N'-methylamide 97%

Supplier: Ambeed
Warning

Synonyms: (S)-TERT-BUTYL 1-(METHOXY(METHYL)AMINO)-1-OXOPROPAN-2-YLCARBAMATE, BOC-L-ALANINE N-METHOXY-N-METHYL AMIDE, (2S)-2-[(tert-butoxy)carbonylamino]-N-methoxy-N-methylpropanamide, Boc-Ala-NMe(OMe)

A234682-100G A234682-10G A234682-250MG A234682-1G A234682-25G A234682-5G
103810-970EA 9 USD
103810-970 103850-726 76976-290 76976-292 76976-294 77089-720 77089-722 77089-724
N-(tert-Butoxycarbonyl)-L-alanine N'-methoxy-N'-methylamide 97%
N-(tert-Butoxycarbonyl)-L-alanine N'-methoxy-N'-methylamide

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₄
MW: 232.28 g/mol
Melting Pt: 148…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00151859
CAS Number: 87694-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR