Boc-Ala-NH2 95%

Supplier: Ambeed
Warning

Synonyms: tert-butyl N-(1-carbamoylethyl)carbamate, Boc-Ala-NH₂, N-Boc-L-alaninamide

A146908-100G A146908-10G A146908-5G A146908-1G A146908-25G
103803-720EA 5 USD
103803-720 103841-814 77137-586 77137-588 77299-552 77317-170 77317-172
Boc-Ala-NH2 95%
Boc-Ala-NH2

Precaución: for Research Use Only

Formula: C₈H₁₆N₂O₃
MW: 188.227 g/mol
Boiling Pt: 343.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD03701447
CAS Number: 85642-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR