N-Boc-3-(4-pyridyl)-L-alanine 95%

Supplier: Ambeed
Warning

Synonyms: Boc-Ala(4-pyridyl)-OH, Boc-(4-Pyridyl)-Ala-OH, Boc-4-Pal-OH

A530707-250MG A530707-10G
103828-566EA 46 USD
103828-566 103839-674 76726-294 77669-354
N-Boc-3-(4-pyridyl)-L-alanine 95%
N-Boc-3-(4-pyridyl)-L-alanine

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O₄
MW: 266.297 g/mol
Boiling Pt: 454.3 °C (760 mm Hg)
Flash Pt: 228.5 °C
Storage Temperature: Freezer
MDL Number: MFCD00672528
CAS Number: 37535-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR