Boc-Aib-OH 97%

Supplier: Ambeed
Warning

Synonyms: BOC-α-Methylalanine, N-Boc-alpha-methylalanine, 2-(Boc-amino)isobutyric Acid, Boc-Aib-OH

A202881-1000G A202881-25G A202881-100G A202881-10G A202881-500G A202881-5G
103809-378EA 351 USD
103809-378 103848-066 77086-180 77086-182 77097-450 77097-452 77097-454 77302-832
Boc-Aib-OH 97%
Boc-Aib-OH

Precaución: for Research Use Only

Formula: C₉H₁₇NO₄
MW: 203.24 g/mol
Boiling Pt: 327.6 °C (760 mm Hg)
Melting Pt: 118…122 °C
Flash Pt: 151.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00042973
CAS Number: 30992-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR