Boc-8-aminooctanoic acid 97%

Supplier: Ambeed
Warning

A128469-10G A128469-250MG A128469-25G A128469-1G A128469-5G A128469-100G
103801-132EA 12 USD
103801-132 76688-554 76688-556 77298-304 77313-450 77313-452 77624-090
Boc-8-aminooctanoic acid 97%
Boc-8-aminooctanoic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₅NO₄
Boiling Pt: 406.0±28.0 °C (760 mm Hg)
Melting Pt: 56…59 °C
Flash Pt: 199.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00270350
CAS Number: 30100-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR