4-((tert-Butoxycarbonyl)amino)benzoic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-4-Abz-OH, 4-(Boc-amino)benzoic Acid
A136088-100G
A136088-10G
A136088-1G
A136088-25G
A136088-5G
103802-270EA
5
USD
103802-270
76719-200
76719-202
77315-162
77315-164
77315-166
4-((tert-Butoxycarbonyl)amino)benzoic acid 98%
4-((tert-Butoxycarbonyl)amino)benzoic acid
Precaución: for Research Use Only
Formula:
C₁₂H₁₅NO₄ MW: 237.3 g/mol Boiling Pt: 336.2 °C (760 mm Hg) Melting Pt: 193 °C Flash Pt: 157.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00037428 CAS Number: 66493-39-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...