Boc-3-Abz-OH 98%

Supplier: Ambeed
Warning

Synonyms: 3-{[(tert-Butoxy)carbonyl]amino}benzoic acid, Boc-3-aminobenzoic acid

A122815-100G A122815-10G A122815-25G A122815-1G A122815-5G
103800-286EA 9 USD
103800-286 77009-512 77009-514 77009-516 77312-406 77312-408
Boc-3-Abz-OH 98%
Boc-3-Abz-OH

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₄
MW: 237.26 g/mol
Boiling Pt: 339.8 °C (760 mm Hg)
Melting Pt: 195 °C
Flash Pt: 159.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00235884
CAS Number: 111331-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR