N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-L-alanine 97%

Supplier: Ambeed
Warning

Synonyms: N-Boc-3-(2-naphthyl)-L-alanine, (S)-alpha-(Boc-amino)-2-naphthalenepropionic Acid, (S)-alpha-tert-Butoxycarbonylamino-2-naphthalenepropionic Acid, Boc-2-Nal-OH

A176946-10G A176946-1G A176946-25G A176946-5G
103807-184EA 16 USD
103807-184 103845-510 76813-406 77082-870 77082-872 77301-478
N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-L-alanine 97%
N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-L-alanine

Precaución: for Research Use Only

Formula: C₁₈H₂₁NO₄
MW: 315.37 g/mol
Boiling Pt: 512.2 °C (760 mm Hg)
Melting Pt: 95 °C
Flash Pt: 263.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00079671
CAS Number: 58438-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR