Bis(4-biphenylyl)amine 97%

Supplier: Ambeed
Warning

Synonyms: Di([1,1'-biphenyl]-4-yl)amine, 4,4'-Iminobis(biphenyl), Bis-biphenyl-4-yl-amine

A115904-100G A115904-10G A115904-1G A115904-25G A115904-5G
103824-508EA 7 USD
103824-508 77284-504 77284-506 77284-508 77284-510 77311-330
Bis(4-biphenylyl)amine 97%
Bis(4-biphenylyl)amine

Precaución: for Research Use Only

Formula: C₂₄H₁₉N
MW: 321.42 g/mol
Melting Pt: 209 °C
Flash Pt: 281.7 °C
Storage Temperature: Ambient
MDL Number: MFCD08276279
CAS Number: 102113-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR