Palladium(II)(pi-cinnamyl) chloride 98%

Supplier: Ambeed
Warning

A432181-1G A432181-5G A432181-250MG A432181-10G
103827-534EA 46 USD
103827-534 103838-600 76842-294 76842-296 76990-508 77568-862
Palladium(II)(pi-cinnamyl) chloride 98%
Palladium(II)(pi-cinnamyl) chloride

Precaución: For research use only.

Formula: C₁₈H₁₈Cl₂Pd₂
Storage Temperature: Refrigerator
MDL Number: MFCD16621470
CAS Number: 12131-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR