Bis[(pinacolato)boryl]methane 95%

Supplier: Ambeed
Warning

A218916-1G A218916-5G A218916-100G A218916-10G A218916-25G
103818-756EA 6 USD
103818-756 103849-630 77088-170 77088-172 77165-154 77165-156 77550-422
Bis[(pinacolato)boryl]methane 95%
Bis[(pinacolato)boryl]methane

Precaución: for Research Use Only

Formula: C₁₃H₂₆B₂O₄
Melting Pt: 48 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD27977747
CAS Number: 78782-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR